Order Entry
Netherlands
ContactUsLinkComponent
Anti-PRD1 Rabbit Monoclonal Antibody [clone: C14A4]
Anti-PRD1 Rabbit Monoclonal Antibody [clone: C14A4]
Catalog # CSTA9115S
Supplier:  Cell Signaling
Anti-PRD1 Rabbit Monoclonal Antibody [clone: C14A4]
Catalog # CSTA9115S
Supplier:  Cell Signaling

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    PRD1
  • Clonality:
    Monoclonal
  • Clone:
    C14A4
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Sequence:
    CYTNDTVPKNANRKYFWRIYSRGEL
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Ambient
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary Antibodies
Antigen: PRD1
Clonality: Monoclonal
Clone: C14A4
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse