Anti-Thymidylate Synthase Rabbit Monoclonal Antibody [clone: D5B3]
Catalog # CSTA9045S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Thymidylate Synthase
- Clonality:Monoclonal
- Clone:D5B3
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Sequence:CKIDDFKAEDFQIEGYNPHPTIK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: Thymidylate Synthase
Clonality: Monoclonal
Clone: D5B3
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human,Mouse,Rat,Monkey