Anti-Catenin-beta (Ser33/Ser37/Thr41) non-phosphate Rabbit Monoclonal Antibody [clone: 'D13A1]
Catalog # CSTA8814S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Catenin-beta (Ser33/Ser37/Thr41) non-phosphate
- Clonality:Monoclonal
- Clone:D13A1
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:LDSGIHSGATTTAPSC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Tested applications:CUT&RUN
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: Catenin-beta (Ser33/Ser37/Thr41) non-phosphate
Clonality: Monoclonal
Clone: 'D13A1
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey