Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: 'D3F9] (Alexa Fluor® 594)
Catalog # CSTA8632S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p38-alpha MAPK (Thr180/Tyr182) phosphate
- Clonality:Monoclonal
- Clone:D3F9
- Conjugation:Alexa Fluor® 594
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:S. cerevisiae,Pig,Monkey,Human,Mink,Rat,Mouse
- Antigen synonyms:p38-delta MAPK (Thr180/Tyr182) phosphate|p38-alpha MAPK (Thr180/Tyr182) phosphate|p38-gamma MAPK (Thr183/Tyr185) phosphate|p38-beta MAPK (Thr180/Tyr182) phosphate
- Sequence:LARHTDDEMtGyVAC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibody Conjugates
Antigen: p38-alpha MAPK (Thr180/Tyr182) phosphate
Clonality: Monoclonal
Clone: 'D3F9
Conjugation: Alexa Fluor 594
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey,Mink,Pig,S. cerevisiae