Anti-Catenin-beta (Ser552) phosphate Rabbit Monoclonal Antibody [clone: 'D8E11]
Catalog # CSTA5651S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Catenin-beta (Ser552) phosphate
- Clonality:Monoclonal
- Clone:D8E11
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CHQDTQRRTsMGG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: Catenin-beta (Ser552) phosphate
Clonality: Monoclonal
Clone: 'D8E11
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse