Anti-PLK Binding Motif (s/tP) phosphate Rabbit Monoclonal Antibody [clone: 'D73F6]
Catalog # CSTA5243S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PLK Binding Motif (s/tP) phosphate
- Clonality:Monoclonal
- Clone:D73F6
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Sequence:CXXXXStPXXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: PLK Binding Motif (s/tP) phosphate
Clonality: Monoclonal
Clone: 'D73F6
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: All