Anti-EIF2S1/eIF2-alpha (Ser52) phosphate Rabbit Monoclonal Antibody [clone: 'D9G8] (Biotin)
Catalog # CSTA5199S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:EIF2S1/eIF2-alpha (Ser52) phosphate
- Clonality:Monoclonal
- Clone:D9G8
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,D. melanogasterm,Rat,Mouse
- Western blot:Yes
- Sequence:GMLLSELsRRRIRSC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: EIF2S1/eIF2-alpha (Ser52) phosphate
Clonality: Monoclonal
Clone: 'D9G8
Conjugation: Biotin
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Monkey,D. melanogasterm