Anti-VEGFR2 (Tyr951) phosphate Rabbit Monoclonal Antibody [clone: '15D2]
Catalogus # CSTA4991S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:VEGFR2 (Tyr951) phosphate
- Clonality:Monoclonal
- Clone:15D2
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:RQGKDyVGAIPVDC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: VEGFR2 (Tyr951) phosphate
Clonality: Monoclonal
Clone: '15D2
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse