Anti-GABA(B)R2 Rabbit Monoclonal Antibody [clone: 'C44A4]
Catalog # CSTA4819S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:GABA(B)R2
- Clonality:Monoclonal
- Clone:C44A4
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGERMGTIKFTQFQDSREVKVGE
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: GABA(B)R2
Clonality: Monoclonal
Clone: 'C44A4
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat