Anti-HIF1A (Pro564) hydroxylate Rabbit Monoclonal Antibody [clone: 'D43B5]
Catalog # CSTA3434S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:HIF1A (Pro564) hydroxylate
- Clonality:Monoclonal
- Clone:D43B5
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Western blot:Yes
- Seq:02
- Environmentally Preferable:
- Sequence:CLEMLA[P-OH]YIPMDD
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: HIF1A (Pro564) hydroxylate
Clonality: Monoclonal
Clone: 'D43B5
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Monkey