Anti-IRS-1 (Ser616) phosphate Rabbit Monoclonal Antibody [clone: 'C15H5]
Catalog # CSTA3203S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:IRS-1 (Ser616) phosphate
- Clonality:Monoclonal
- Clone:C15H5
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:YMPMsPGVAPVPSGRC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: IRS-1 (Ser616) phosphate
Clonality: Monoclonal
Clone: 'C15H5
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat