Anti-ATR (Thr1989) phosphate Rabbit Monoclonal Antibody [clone: 'D5K8W]
CSTA30632S
- Antibody type:Primary
- Antigen symbol:ATR (Thr1989) phosphate
- Clonality:Monoclonal
- Clone:D5K8W
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Sequence:ENEtPPEGC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: ATR (Thr1989) phosphate
Clonality: Monoclonal
Clone: 'D5K8W
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human