Anti-LEF1 Rabbit Monoclonal Antibody [clone: C12A5]
Catalog # CSTA2230S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:LEF1
- Clonality:Monoclonal
- Clone:C12A5
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CGHEVARQAQTSQEPYHDKAREHPDDGK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: LEF1
Clonality: Monoclonal
Clone: C12A5
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat