Anti-Akt-phosphate Substrate (Ser/Thr) phosphate Rabbit Monoclonal Antibody [clone: 110B7E] (Sepharose Bead)
Catalog # CSTA9646S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Akt-phosphate Substrate (Ser/Thr) phosphate
- Clonality:Monoclonal
- Clone:110B7E
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Sequence:CXXXRXRXXtXXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: Akt-phosphate Substrate (Ser/Thr) phosphate
Clonality: Monoclonal
Clone: 110B7E
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: