Anti-ATM Substrate motif [L(s/t)Q] phosphate Rabbit Monoclonal Antibody [clone: D23H2/D69H5) MultiMab™ Rabbit mAb mix]
Catalog # CSTA9607S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:ATM Substrate motif [L(s/t)Q] phosphate
- Clonality:Monoclonal
- Clone:D23H2/D69H5) MultiMab™ Rabbit mAb mix
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Sequence:CXXXXXX(s/t)QXXXXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ATM Substrate motif [L(s/t)Q] phosphate
Clonality: Monoclonal
Clone: D23H2/D69H5) MultiMab™ Rabbit mAb mix
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species