Anti-Catenin-beta (Ser37/Thr41/Ser45) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA9564S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Catenin-beta (Ser37/Thr41/Ser45) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Sequence:GIHsGATtTAPsLSGKGC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Catenin-beta (Ser37/Thr41/Ser45) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human