Anti-GSK3-alpha (Ser21) phosphate Rabbit Monoclonal Antibody [clone: 37F11]
Type: Primary
Antigen: GSK3-alpha (Ser21) phosphate
Clonality: Monoclonal
Clone: 37F11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
- Catalog No:
- CSTA9327S
- CSTA9327L
- Antigen symbol:
- GSK3-alpha (Ser21) phosphate
- GSK3-alpha (Ser21) phosphate
- Antigen synonyms:
- GSK3-beta (Ser9) phosphate
- GSK3-beta (Ser9) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 37F11
- 37F11
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CRARTSsFAEPGG
- CRARTSsFAEPGG
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Cold
- Cold
Frequently Bought Together