Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: 3D7]
Type: Primary
Antigen: p38-alpha MAPK (Thr180/Tyr182) phosphate
Clonality: Monoclonal
Clone: 3D7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;D. melanogaster;Pig;S. cerevisiae
- Catalog No:
- CSTA9215S
- CSTA9215L
- Antigen symbol:
- p38-alpha MAPK (Thr180/Tyr182) phosphate
- p38-alpha MAPK (Thr180/Tyr182) phosphate
- Antigen synonyms:
- p38-delta MAPK (Thr180/Tyr182) phosphate|p38-alpha MAPK (Thr180/Tyr182) phosphate|p38-gamma MAPK (Thr183/Tyr185) phosphate|p38-beta MAPK (Thr180/Tyr182) phosphate
- p38-delta MAPK (Thr180/Tyr182) phosphate|p38-alpha MAPK (Thr180/Tyr182) phosphate|p38-gamma MAPK (Thr183/Tyr185) phosphate|p38-beta MAPK (Thr180/Tyr182) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 3D7
- 3D7
- Reactivity:
- S. cerevisiae, Pig, Monkey, Human, D. melanogasterm, Rat, Mouse
- S. cerevisiae, Pig, Monkey, Human, D. melanogasterm, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- LARHTDDEMtGyVAC
- LARHTDDEMtGyVAC
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Cold
- Cold
Frequently Bought Together