Anti-HP1-beta Rabbit Monoclonal Antibody [clone: D2F2]
CSTA8676S
- Antibody type:Primary
- Antigen symbol:HP1-beta
- Clonality:Monoclonal
- Clone:D2F2
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- ChIP:Yes
- Sequence:CPDLIAEFLQSQKTAHETDK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Tested applications:CUT&RUN
- Pk:100 µl
Type: Primary
Antigen: HP1-beta
Clonality: Monoclonal
Clone: D2F2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey