Anti-H4 (Lys5) acetylate Rabbit Monoclonal Antibody [clone: D12B3]
CSTA8647S
- Antibody type:Primary
- Antigen symbol:H4 (Lys5) acetylate
- Clonality:Monoclonal
- Clone:D12B3
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:SGRGkGGKGLGC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: H4 (Lys5) acetylate
Clonality: Monoclonal
Clone: D12B3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey