Anti-Vimentin (Ser56) phosphate Rabbit Monoclonal Antibody [clone: D5H2]
Catalog # CSTA7391S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Vimentin (Ser56) phosphate
- Clonality:Monoclonal
- Clone:D5H2
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CSRSLYASsPGGVY
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Vimentin (Ser56) phosphate
Clonality: Monoclonal
Clone: D5H2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey