Anti-SRC-3 Rabbit Monoclonal Antibody [clone: D1F11]
CSTA5765S
- Antibody type:Primary
- Antigen symbol:SRC-3
- Clonality:Monoclonal
- Clone:D1F11
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Sequence:REQNGYRPNPNPVGQGIRPPMAGC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: SRC-3
Clonality: Monoclonal
Clone: D1F11
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Monkey