Anti-beta-Actin Rabbit Monoclonal Antibody [clone: 13E5] (HRP)
Catalog # CSTA5125S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:beta-Actin
- Clonality:Monoclonal
- Clone:13E5
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Sequence:DDDIAALVVDNGSGC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: beta-Actin
Clonality: Monoclonal
Clone: 13E5
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;Bovine;Pig