Anti-SAPK/JNK1 (Thr183/Tyr185) phosphate Rabbit Monoclonal Antibody [clone: 81E11] (Sepharose Bead)
CSTA4306S
- Antibody type:Primary
- Antigen symbol:SAPK/JNK1 (Thr183/Tyr185) phosphate
- Clonality:Monoclonal
- Clone:81E11
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:S. cerevisiae,Human,D. melanogasterm,Rat,Mouse
- Sequence:SFMMtPyVVTRYYRC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:400 µl
Type: Primary
Antigen: SAPK/JNK1 (Thr183/Tyr185) phosphate
Clonality: Monoclonal
Clone: 81E11
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;D. melanogaster;S. cerevisiae