Anti-p38-alpha MAPK (Thr180/Tyr182) phosphate Rabbit Monoclonal Antibody [clone: 3D7] (Biotinylated)
Catalog # CSTA4092S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p38-alpha MAPK (Thr180/Tyr182) phosphate
- Clonality:Monoclonal
- Clone:3D7
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:S. cerevisiae,Pig,Monkey,Human,D. melanogasterm,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:p38-delta MAPK (Thr180/Tyr182) phosphate|p38-alpha MAPK (Thr180/Tyr182) phosphate|p38-gamma MAPK (Thr183/Tyr185) phosphate|p38-beta MAPK (Thr180/Tyr182) phosphate
- Sequence:LARHTDDEMtGyVAC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p38-alpha MAPK (Thr180/Tyr182) phosphate
Clonality: Monoclonal
Clone: 3D7
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;D. melanogaster;Pig;S. cerevisiae