Anti-Drosha Rabbit Monoclonal Antibody [clone: D30F3]
Catalog # CSTA3410S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Drosha
- Clonality:Monoclonal
- Clone:D30F3
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:CSAKARRTGIRHSIYPGEEAIK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Drosha
Clonality: Monoclonal
Clone: D30F3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse