Anti-CSF1R (Tyr809) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA3154S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:CSF1R (Tyr809) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Sequence:CDIMNDSNy(V/I)VKGNA
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CSF1R (Tyr809) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse