Anti-BCL-xL Rabbit Monoclonal Antibody [clone: 54H6] (Alexa Fluor 488)
Catalog # CSTA2767S
Leverancier: Cell Signaling
Specificaties
- Antibody type:Primary
- Antigen symbol:BCL-xL
- Clonality:Monoclonal
- Clone:54H6
- Conjugation:Alexa Fluor® 488
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Sequence:CINGNPSWHLAD
- Storage temperature:4 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:50 Tests
Specificaties
Over dit artikel
Type: Primary
Antigen: BCL-xL
Clonality: Monoclonal
Clone: 54H6
Conjugation: Alexa Fluor 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey