Anti-p38-alpha MAPK Rabbit Monoclonal Antibody [clone: D13E1] (Biotinylated)
Catalog # CSTA2387S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p38-alpha MAPK
- Clonality:Monoclonal
- Clone:D13E1
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Hamster,Pig,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:p38-beta MAPK|p38-alpha MAPK|p38-gamma MAPK
- Sequence:TYDEVISFVPPPLDQEEMESC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p38-alpha MAPK
Clonality: Monoclonal
Clone: D13E1
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: House;Mouse;Rat;Hamster;Monkey;Bovine;Pig