Gas Welding Goggles
Catalog # PRTWPW60BGR
Supplier: Portwest
Specifications
- Colour:Green
- Description:Gas welding goggles, indirect ventilation
- Pk:1
Specifications
About this item
When using oxyacetylene for gas cutting or welding, the PW60 goggles offers high performance with a shade factor five.
- Indirect ventilation
- Flexible PVC frame for ultra comfort
- Stand-off position for intermittent use
- Adjustable elastic headband for a perfect fit
- Anti-scratch coating for added durability
- 99% UV protection
- Retail bag aids presentation for retail sales
A soft comfortable frame with flip up lens allows the goggle to be used for grinding operations. Indirect ventilation goggles are individually packaged.
Certifications: EN 169 (shade 5).