Anti-NDRG1 (Thr346) phosphate Rabbit Monoclonal Antibody [clone: D98G11] (HRP)
CSTA8947S
- Antibody type:Primary
- Antigen symbol:NDRG1 (Thr346) phosphate
- Clonality:Monoclonal
- Clone:D98G11
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CEGTRSRSHtSEG
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: NDRG1 (Thr346) phosphate
Clonality: Monoclonal
Clone: D98G11
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey