Anti-PARP1 (Asp214, 89 kD) cleaved Rabbit Monoclonal Antibody [clone: D64E10] (Sepharose Bead)
Catalog # CSTA5838S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:PARP1 (Asp214, 89 kD) cleaved
- Clonality:Monoclonal
- Clone:D64E10
- Conjugation:Sepharose
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human
- Sequence:GVDEVAKKKSKKEKDKDSC
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PARP1 (Asp214, 89 kD) cleaved
Clonality: Monoclonal
Clone: D64E10
Conjugation: Sepharose Bead
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Monkey