Anti-SHIP2 (Tyr1135) phosphate Rabbit Monoclonal Antibody [clone: D33C6]
CSTA5445S
- Type antilichaam:Primair
- antigeen symbool:SHIP2 (Tyr1135) phosphate
- Clonality:Monoklonaal
- Clone:D33C6
- Flowcytometrie:Yes
- Host:Rabbit
- ImmunoNeerslag:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Sequentie:CTLSEVDyAPAGPAR
- Opslagtemperatuur:−20 °C
- Shipping temperature:Ambient
- VPE:100 µl
Type: Primary
Antigen: SHIP2 (Tyr1135) phosphate
Clonality: Monoclonal
Clone: D33C6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human