Anti-STAT3 (Tyr705) phosphate Rabbit Monoclonal Antibody [clone: D3A7] (Alexa Fluor 647)
Catalog # CSTA4324S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:STAT3 (Tyr705) phosphate
- Clonality:Monoclonal
- Clone:D3A7
- Conjugation:Alexa Fluor® 647
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Sequence:ADPGSAAPyLKTKFIC
- Storage temperature:4 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:50 Tests
Specifications
About this item
Type: Primary
Antigen: STAT3 (Tyr705) phosphate
Clonality: Monoclonal
Clone: D3A7
Conjugation: Alexa Fluor 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey