Anti-ATM Substrate (Thr) phosphate Rabbit Monoclonal Antibody [clone: 4F7]
Catalog # CSTA2909S
Leverancier: Cell Signaling
Specificaties
- Antibody type:Primary
- Antigen symbol:ATM Substrate (Thr) phosphate
- Clonality:Monoclonal
- Clone:4F7
- Host:Rabbit
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- Sequence:CXXXXXXLtQ[D/E]XXXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specificaties
Over dit artikel
Type: Primary
Antigen: ATM Substrate (Thr) phosphate
Clonality: Monoclonal
Clone: 4F7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Any species