Anti-AMPK-alpha1 (Ser496) phosphate Rabbit Monoclonal Antibody [clone: 45F5]
Catalog # CSTA2537S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:AMPK-alpha1 (Ser496) phosphate
- Clonality:Monoclonal
- Clone:45F5
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:TPQRSGsVSNYRSC
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: AMPK-alpha1 (Ser496) phosphate
Clonality: Monoclonal
Clone: 45F5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey