Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

3,3',5,5'-Tetramethylbenzidine (TMB), research grade

Marketsource
Supplier:   Serva
3,3',5,5'-Tetramethylbenzidine (TMB), research grade
Supplier:  Serva
Marketsource
Certificates
3,3',5,5'-Tetramethylbenzidine (TMB), research grade

2 Options Available Below

Product Details & Documents

Substrate for horseradish peroxidase.

More sensitive than ABTS and more stable and less toxic than diaminobenzidine. See also TMB Ready-To-Use ELISA Substrate (cat. no. 37068) and SERVAColor TMB Blot Solution (cat. no. 37071).

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Boiling Pt:402 °C (1013 hPa)
  • Melting Pt:166…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 83,00 €
    83,00 €
    1 * 5 g
       83,00 €

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 311,00 €
    311,00 €
    1 * 25 g
       311,00 €

    Specifications:
    Marketsource
    Marketsource