Sodium cholate
Marketsource
Leverancier: Serva
Jump to:
About this item
Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
- CAS Number:361-09-1
- Formula:C₂₄H₃₉NaO₅
- MDL Number:MFCD00150749
- Molecular Weight:430.56 g/mol
- Melting Pt:198 °C
- Storage Temperature:Ambient
- EINECS:206-643-5
2 Options Available Below
Product Details & Documents
Synonyms: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
- CAS Number:361-09-1
- Formula:C₂₄H₃₉NaO₅
- MDL Number:MFCD00150749
- Molecular Weight:430.56 g/mol
- Melting Pt:198 °C
- Storage Temperature:Ambient
- EINECS:206-643-5
Product Family Options
Product Information
- Pack typePkBeschikbaarheidPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



