Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
Cinnamaldehyde
Catalog # ROTH5549.1
Supplier:  Roth Carl
CAS Number:  
Irritant
Cinnamaldehyde
Catalog # ROTH5549.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL

Specifications

About this item

 :
 :132.16 g/mol
Merck Index:12,02356
Boiling Pt:251 °C (1013 hPa)
Melting Pt:–8 °C
Density:1.112 g/cm³ (20 °C)
Flash Pt:138 °C
Storage Temperature:Ambient
EINECS:203-213-9
 :GHS07
CAS Number:104-55-2
Formula:C₉H₈O
MDL Number:MFCD00007000