Order Entry
Netherlands
ContactUsLinkComponent
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Health
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Catalog # ROTH5028.1
Supplier:  Roth Carl
CAS Number:  
Health
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Catalog # ROTH5028.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :505.7 g/mol
Melting Pt:227…229 °C
EINECS:205-676-2
 :GHS08
Storage Temperature:Refrigerator
CAS Number:146-68-9
Formula:C₁₉H₁₃ClIN₅O₂
MDL Number:MFCD00011961