Aluminium chloride Reag. Ph. Eur. 1002702
Catalog # 87765.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:133.34 g/mol
Storage Temperature:Ambient
UN:3286
ADR:3 (6.1, 8),I
CAS Number:7446-70-0
Formula:AlCl₃
MDL Number:MFCD00003422