Order Entry
Netherlands
ContactUsLinkComponent
Nitric acid 100 ml/l
Nitric acid 100 ml/l
Catalog # PRLSMC310519.1000
Supplier:  PROLABO CMP
CAS Number:  
Nitric acid 100 ml/l
Catalog # PRLSMC310519.1000
Supplier:  PROLABO CMP
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349