Order Entry
Netherlands
ContactUsLinkComponent
4-Bromopyridinium chloride
4-Bromopyridinium chloride
Catalog # MOLE65801625-5G
Supplier:  Molekula
CAS Number:  
4-Bromopyridinium chloride
Catalog # MOLE65801625-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :194.46 g/mol
Melting Pt:235…238 °C
EINECS:243-128-4
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:19524-06-2
Formula:C₅H₅BrClN
MDL Number:MFCD00012828