
(3-Chloropropyl)trichlorosilane ≥98.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2550-06-3
:
Formula:ClH₂CCH₂CH₂SiCl₃
MDL Number:MFCD00013694
:211.98 g/mol
Boiling Pt:176…177 °C (1013 hPa)
Density:1.359 g/cm³ (20 °C)
EINECS:219-844-8
UN:2987
ADR:8,II
:M-Clarity™ quality level = MQ200
:GHS05
Specification Test Results
- Assay (GC, area%):≥ 98.0 % (a/a)
- Density (d 20 °C/ 4 °C):1.358 - 1.362
- Identity (IR):passes test
- Flash Point:80 °C
- Density:1.360 g/cm3 (20 °C)
Specification Test Results
Frequently Bought Together