
N-Isopropylbenzylamine for synthesis, Sigma-Aldrich®
Catalog # 8.41618.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Molar Mass:149.24 g/mol
- Boiling Point:199 - 200 °C (1013 hPa)
- Flash Point:87 °C
- Density:0.91 g/cm3 (20 °C)
Specification Test Results
About this item
:
:149.24 g/mol
Boiling Pt:200 °C (1013 hPa)
Melting Pt:∼ 0 °C
Density:0.89 g/cm³ (20 °C)
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:203-067-6
UN:2735
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:102-97-6
Formula:C₆H₅CH₂N(CH₃)(CH₂CH₃)
MDL Number:MFCD00008863