Order Entry
Netherlands
ContactUsLinkComponent
(Methoxymethyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®
Irritant
Environment
(Methoxymethyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.18580.0050
Supplier:  Merck
CAS Number:  
Irritant
Environment
(Methoxymethyl)triphenylphosphonium chloride for synthesis, Sigma-Aldrich®
Catalog # 8.18580.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble,(decomposition)
  • Melting Point:
    202 °C
  • Molar Mass:
    342.8 g/mol
  • Bulk Density:
    450 - 500 kg/m3
  • Boiling Point:
    413 °C
  • Vapor Pressure:
     - 1 hPa (20 °C)
  • Flash Point:
    250 °C
  • Hazard Class:
    9
  • Hazard Class Description:
    Miscellaneous Hazardous Material

Specification Test Results

About this item

 :
 :342.8 g/mol
Melting Pt:190 °C
Storage Temperature:Ambient
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
EINECS:223-664-5
 :GHS07
 :GHS09
CAS Number:4009-98-7
Formula:C₂₀H₂₀ClOP
MDL Number:MFCD00011800