


Bis(2-chloroethyl)ether, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-44-4
:
Formula:C₄H₈Cl₂O
MDL Number:MFCD00000975
:143.01 g/mol
Merck Index:13,03091
Boiling Pt:178 °C (1013 hPa)
Melting Pt:–44 °C
Density:1.22 g/cm³ (20 °C)
EINECS:203-870-1
UN:1916
ADR:6.1 (3),II
:M-Clarity™ quality level = MQ200
Flash Pt:55 °C
:GHS02
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Solubility:0.01 g/l (20 °C)
- Melting Point:-51.9 °C
- Molar Mass:143.01 g/mol
- Boiling Point:177 - 178 °C
- Vapor Pressure:1.07 hPa (20 °C)
- Flash Point:55 °C
- Refractive Index:1.457 (20 °C, 589 nm)
- Explosion Limit:0.8 %(V)
- Density:1.22 g/cm3 (20 °C)
- Ignition Point:365 °C
Specification Test Results
Frequently Bought Together