Order Entry
Netherlands
ContactUsLinkComponent
 
Irritant
1-Naphtholbenzeine Reag. Ph. Eur., Millipore®
Catalog # 1.06202.0005
Supplier:  Merck
CAS Number:  
Irritant
1-Naphtholbenzeine Reag. Ph. Eur., Millipore®
Catalog # 1.06202.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    243 - 245 °C
  • Molar Mass:
    374.44 g/mol
  • Bulk Density:
    120 - 300 kg/m3

Specification Test Results

About this item

 :
 :374.44 g/mol
Melting Pt:243…245 °C
 :M-Clarity™ quality level = MQ200
EINECS:205-656-3
 :GHS07
Storage Temperature:Ambient
CAS Number:145-50-6
 :6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492