Order Entry
Netherlands
ContactUsLinkComponent
Organic reference standard, 4-Nitrotoluene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Organic reference standard, 4-Nitrotoluene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL15615400AL
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 4-Nitrotoluene 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERL15615400AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    99-99-0
  • Description:
    4-Nitrotoluene 10 µg/ml in acetonitrile
  • Formula:
    C₇H₇NO₂
  • Description:
    4-Nitrotoluene

Specifications

About this item