Order Entry
Netherlands
ContactUsLinkComponent
6-Methoxy-m-toluidine
Health
Irritant
6-Methoxy-m-toluidine
Catalog # EHERC15081000
Supplier:  EHRENSTORFER
CAS Number:  
Health
Irritant
6-Methoxy-m-toluidine
Catalog # EHERC15081000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :137.18 g/mol
Boiling Pt:235 °C (1013 hPa)
Boiling Pt:235 °C (1013 hPa)
Melting Pt:55.5 °C
Melting Pt:55,5 °C
Density:1 g/cm³ (20 °C)
EINECS:204-419-1
 :GHS08
 :GHS07
CAS Number:120-71-8
Formula:C₈H₁₁NO
MDL Number:MFCD00007815